missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO42 Monoclonal Antibody (OTI1C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800209
This item is not returnable.
View return policy
Description
Members of the F-box protein family, such as FBXO42, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.Specifications
| FBXO42 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6P3S6 | |
| FBXO42 | |
| Human recombit protein fragment corresponding to amino acids 356-717 of human FBXO42 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1C1 | |
| Unconjugated | |
| FBXO42 | |
| Fbx42, JFK | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54455 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction