missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO42 Monoclonal Antibody (OTI1C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800209S
This item is not returnable.
View return policy
Description
FBXO42 Monoclonal antibody specifically detects FBXO42 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| FBXO42 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q93062 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11030 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1C1 | |
| Unconjugated | |
| Rbpms | |
| 6720460I06Rik; F-box only protein 42; F-box protein 42; Fbx42; Fbxo42; JFK; just one F-box and Kelch domain-containing protein; KIAA1332; LOC527807 protein; mKIAA1332 | |
| Full length human recombinant protein of human RBPMS produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction