missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO44 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-467IR
This item is not returnable.
View return policy
Description
FBXO44 Polyclonal antibody specifically detects FBXO44 in Human samples. It is validated for Western Blot
Specifications
| FBXO44 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 93611 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| FBG3 DKFZp781J0852, F-box gene 3, F-box only protein 44, F-box protein 44, F-box protein FBX30, F-box/G-domain protein 3, FBX30FBX6AFbx44, FBX44, Fbxo6a, MGC14140 | |
| A synthetic peptide containing residues 103-119 EWKVEDLSRDQRKEFPN) of the human FBG3 protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction