missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA123511
This item is not returnable.
View return policy
Description
PA1-23511 detects FBG3 (F Box 3) in Human samples.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs family, similar to F-box only protein 2 and F-box only protein 6. Four alternatively spliced transcript variants encoding two distinct isoforms have been found for this gene.
Specifications
| FBXO44 | |
| Polyclonal | |
| Unconjugated | |
| FBXO44 | |
| 5730411K09; AV001623; DKFZp781J0852; F Box Gama 3; FBG3; F-box gene 3; F-box only protein 44; F-box only protein 6a; F-box protein 44; F-box protein FBX30; F-box/G-domain protein 3; FBX30; Fbx44; FBX6A; FBXO44; FBXO6A; MGC14140; RGD1562463; RP3-330O12.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 93611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9H4M3 | |
| FBXO44 | |
| Residues 103-119 (EWKVEDLSRDQRKEFPN) of the human FBXO44 protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido