Learn More
FBXO6 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98796PCP
Product Code. 18634825
  • € 407.00 / 0.10mL
Description
FBXO6 Polyclonal antibody specifically detects FBXO6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FBXO6
Polyclonal
FBG2F-box/G-domain protein 2, F-box only protein 6, F-box protein 6, F-box protein Fbx6, F-box protein that recognizes sugar chains 2, Fbx6b, FBX6FBS2F-box protein FBG2