missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXO6 Monoclonal Antibody (OTI7A10)
GREENER_CHOICE

Product Code. 15996874
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15996874 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15996874 Supplier Invitrogen™ Supplier No. MA527288

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FBXO6 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42 kDa) which may be involved in the control of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FBXO6
Gene Accession No. Q9NRD1
Gene Alias AA408845; FBG1; Fbg2; F-box only protein 6; F-box only protein 6 b; F-box only protein 6b; F-box protein 6; F-box protein FBG2; F-box protein Fbx6; F-box protein that recognizes sugar chains 2; F-box/G-domain protein 2; Fbs1; FBS2; FBX2; FBX6; Fbx6b; FBXO6; Fbxo6b; NFB42; RP3-330O12.3
Gene Symbols FBXO6
Host Species Mouse
Immunogen Full length human recombinant protein of FBXO6 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26270
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.