missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO6 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67697-1-IG-150UL
This item is not returnable.
View return policy
Description
The FBXO6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBXO6. This antibody recognizes human, mouse, rat antigen. The FBXO6 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| FBXO6 | |
| Monoclonal | |
| 1200 μg/mL | |
| FBG2; FBS2; FBX6; Fbx6b | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 34 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 3B7F9 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 26270 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction