missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111328
This item is not returnable.
View return policy
Description
Immunogen sequence: RVTNSSIVVS PKMTRNQASS EAQPGQKHGQ EEAAQSPYRA VVQI Highest antigen sequence indentity to the following orthologs: Rat - 33%, Mouse - 32%.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42 kDa) which may be involved in the control of the cell cycle.
Specifications
| FBXO6 | |
| Polyclonal | |
| Unconjugated | |
| FBXO6 | |
| AA408845; FBG1; Fbg2; F-box only protein 6; F-box only protein 6 b; F-box only protein 6b; F-box protein 6; F-box protein FBG2; F-box protein Fbx6; F-box protein that recognizes sugar chains 2; F-box/G-domain protein 2; Fbs1; FBS2; FBX2; FBX6; Fbx6b; FBXO6; Fbxo6b; NFB42; RP3-330O12.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NRD1 | |
| FBXO6 | |
| Recombinant Protein corresonding to Human FBXO6. Recombinant protein control fragment (Product #RP-101230). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction