missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 572.00
Specifications
| Antigen | FBXO7 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18219262
|
Novus Biologicals
NBP2-58563 |
100 μL |
€ 572.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674359
|
Novus Biologicals
NBP2-58563-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FBXO7 Polyclonal specifically detects FBXO7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| FBXO7 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 25793 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PTLGHLRSHLRQSLLCTWGYSSNTRFTITLNYKDPLTGDEETLASYGIVSGDLICLILQDDIPAPNIPSSTDSEHSSLQNNEQPSLA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| F-box only protein 7, F-box protein 7, FBX, FBX07, FBX7Fbx, PARK15DKFZp686B08113, PKPS | |
| FBXO7 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title