missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38432-100ul
This item is not returnable.
View return policy
Description
FBXO7 Polyclonal antibody specifically detects FBXO7 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| FBXO7 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| F-box only protein 7, F-box protein 7, FBX, FBX07, FBX7Fbx, PARK15DKFZp686B08113, PKPS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 353-522 of human FBXO7 (NP_036311.3).,, Sequence:, LSLSAVCRDLFTASNDPLLWRFLYLRDFRDNTVRVQDTDWKELYRKRHIQRKESPKGRFVMLLPSSTHTIPFYPNPLHPRPFPSSRLPPGIIGGEYDQRPTLPYVGDPISSLIPGPGETPSQFPPLRPRFDPVGPLPGPNPILPGRGGPNDRFPFRPSRGRPTDGRLSFM | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25793 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction