missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16452
This item is not returnable.
View return policy
Description
FBXO7 Polyclonal specifically detects FBXO7 in Human samples. It is validated for Western Blot.
Specifications
| FBXO7 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| F-box only protein 7, F-box protein 7, FBX, FBX07, FBX7Fbx, PARK15DKFZp686B08113, PKPS | |
| Rabbit | |
| 59 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Pig (83%), Rabbit (80%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9Y3I1 | |
| FBXO7 | |
| Recombinant protein encompassing a sequence within the center region of human FBXO7. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 25793 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction