missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXO7 Polyclonal Antibody

Product Code. 15987884
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15987884 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15987884 Supplier Invitrogen™ Supplier No. PA565118

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PTLGHLRSHLR QSLLCTWGYS SNTRFTITLN YKDPLTGDEE TLASYGIVSG DLICLILQDD IPAPNIPSST DSEHSSLQNN EQPSLA Highest antigen sequence identity to the following orthologs - mouse 79%, rat 79%.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it may play a role in regulation of hematopoiesis. Alternatively spliced transcript variants of this gene have been identified with the full-length natures of only some variants being determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXO7
Gene Accession No. Q9Y3I1
Gene Alias 2410015K21Rik; A230052G17Rik; AI117679; AW260462; C85118; DKFZp686B08113; F-box only protein 7; F-box protein 7; FBX; FBX07; FBX7; FBXO7; hypothetical protein LOC558097; PARK15; PKPS; si:ch211-286m4.2; Unknown (protein for MGC:134509); zgc:114096
Gene Symbols FBXO7
Host Species Rabbit
Immunogen Recombinant Human FBXO7. Recombinant protein control fragment (Product #RP-104642).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.