Learn More
FBXO8 Mouse anti-Human, Clone: OTI7E12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807552
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It contains a C-terminal amino acid sequence that bears a significant similarity with a portion of yeast Sec7p, a critical regulator of vesicular protein transport. This human protein may interact with ADP-ribosylation factor(s)(ARFs) and exhibit ARF-GEF (guanine nucleotide exchange factor) activity.Specifications
| FBXO8 | |
| Monoclonal | |
| Unconjugated | |
| FBXO8 | |
| DC10, FBS, FBX8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26269 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI7E12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NRD0 | |
| FBXO8 | |
| Human recombit protein fragment corresponding to amino acids 1-231 of human FBXO8 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.