missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO9, Mouse anti-Human, Clone: 1G12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00026268-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: HYRLSQDTDNQTKVFAVITKKKEEKPLDYKYRYFRRVPVQEADQSFHVGLQLCSSGHQRFNKLIWIHHSCHITYKSTGETAVSAFEIDKMYTPLFFARVRSYTAFSERPLSpecifications
| FBXO9 | |
| Monoclonal | |
| Unconjugated | |
| F-box protein 9 | |
| DKFZp434C0118/FBX9/KIAA0936/NY-REN-57/VCIA1/dJ341E18.2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1G12 | |
| In 1x PBS, pH 7.4 | |
| NM_012347 | |
| FBXO9 | |
| FBXO9 (NP_036479, 338 a.a. ∼ 447 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 26268 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction