missing translation for 'onlineSavingsMsg'
Learn More

FBXW7 Monoclonal Antibody (OTI13H4), TrueMAB™, OriGene

Product Code. p-200015451
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17511773 30 μL 30µL
17591763 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17511773 Supplier Origene Technologies Supplier No. TA802934S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FBXW7 Monoclonal antibody specifically detects FBXW7 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen FBXW7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI13H4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KRT7
Gene Accession No. P08729
Gene Alias 1110001A17Rik; AGO; archipelago; Archipelago homolog; CDC4; DKFZp686F23254; F-box and WD repeat domain containing 7; F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase; F-box and WD-40 domain protein 7; F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila); F-box and WD-40 domain protein 7, archipelago homolog; F-box and WD-40 domain-containing protein 7; f-box only protein 30; F-box protein FBW7; F-box protein FBX30; F-box protein Fbxw6; F-box protein SEL-10; F-box/WD repeat-containing protein 7; F-box-WD40 repeat protein 6; FBW6; FBW7; Fbwd6; FBX30; Fbxo30; FBXW6; FBXW7; FLJ11071; FLJ16457; hAgo; hCdc4; homolog of C elegans sel-10; SEL10; SEL-10
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-114 and 340-469 of human KRT7 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.