Learn More
FBXW7 Monoclonal Antibody (OTI2E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802858S
Encompass_Preferred
 View more versions of this product
Product Code. 17501633
  • € 242.00 / 30µL
Description
FBXW7 Monoclonal antibody specifically detects FBXW7 in Human samples. It is validated for Western Blot
Specifications
Specifications
FBXW7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P78368
Mouse
Affinity chromatography
RUO
1455
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI2E12
Unconjugated
Csnk1g2
1110001A17Rik; AGO; archipelago; Archipelago homolog; CDC4; DKFZp686F23254; F-box and WD repeat domain containing 7; F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase; F-box and WD-40 domain protein 7; F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila); F-box and WD-40 domain protein 7, archipelago homolog; F-box and WD-40 domain-containing protein 7; f-box only protein 30; F-box protein FBW7; F-box protein FBX30; F-box protein Fbxw6; F-box protein SEL-10; F-box/WD repeat-containing protein 7; F-box-WD40 repeat protein 6; FBW6; FBW7; Fbwd6; FBX30; Fbxo30; FBXW6; FBXW7; FLJ11071; FLJ16457; hAgo; hCdc4; homolog of C elegans sel-10; SEL10; SEL-10
Human recombinant protein fragment corresponding to amino acids 163-415 of human CSNK1G2 produced in E.coli.