missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXW9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 307.65
Specifications
| Antigen | FBXW9 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600732
|
Novus Biologicals
NBP2-97608-100ul |
100 μg |
€ 326.00 € 307.65 / 100µL Save € 18.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609264
|
Novus Biologicals
NBP2-97608-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FBXW9 Polyclonal antibody specifically detects FBXW9 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| FBXW9 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| F-box and WD repeat domain containing 9, F-box and WD-40 domain protein 9, F-box and WD-40 domain-containing protein 9, F-box and WD-40 repeat containing protein 9, specificity factor for SCF ubiquitin ligase | |
| Produced in rabbits immunized with E. coli-derived Human FBXW9 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 84261 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title