missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ FC Beads V450 (RUO)
Description
- Fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD™ flow cytometers.
- The combination of BD™ CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels of the instrument.
- BD FC beads can also be used to standardize multiple instruments.
Specifications
Specifications
| Content And Storage | Store tubes at 2°C–8°C in the foil pouch. The tubes should not be frozen. Protect the tubes from exposure to light and humidity. |
| Format | V450 |
| No. of Tests | 5 Tests |
| Particle Size | 3 μm |
| Quantity | 5 Tests |
| Stability | The beads and dilution buffer are stable until the expiration date shown on the pouch and bottle labels when stored as directed. Do not use after the expiration date. |
| Product Type | Fluorescent Polystyrene Beads |
Safety and Handling
Product Identifier
- BD FC Beads V450
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: 2-methylisothiazol-3(2H)-one
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?