missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc epsilon RI alpha Antibody (773735), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB66781D
This item is not returnable.
View return policy
Description
Fc epsilon RI alpha Monoclonal antibody specifically detects Fc epsilon RI alpha in Human samples. It is validated for Serum
Specifications
| Fc epsilon RI alpha | |
| Monoclonal | |
| DyLight 350 | |
| FCE1A,fcepsilonri,FCER1A,FcERI,Fcr-5 | |
| Mouse myeloma cell line NS0-derived recombinant human Fc epsilon RI alpha , Val26-Gln205, Accession # P12319 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 773735 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction