missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc epsilon RI Antibody (9E1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63269APC
This item is not returnable.
View return policy
Description
Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Fc epsilon RI | |
| Monoclonal | |
| APC | |
| alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
| Made to Human Fc EPSILON R1 ALPHA | |
| 0.1 mL | |
| Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
| 2205 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 9E1 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction