missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RI/CD64 Antibody (008), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90404CL1
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Mouse samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Fc gamma RI/CD64 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fc gamma RI/CD64 (Accession#: NP_034316.1; Met 1-Pro 297). | |
| 0.1 mL | |
| CD Markers, Immunology | |
| 2209 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 008 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction