missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RI/CD64 Antibody (276426) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB12571M
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| Fc gamma RI/CD64 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 276426 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RI, Gln16-Pro288, Accession # P12314.2 | |
| 0.1 mL | |
| CD Markers, Immunology | |
| 2209 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction