Learn More
Novus Biologicals™ Fc gamma RI/CD64 Antibody (Medarex Patent Anti-Fc-Gamma-R1), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28132-50ug
 View more versions of this product
Product Code. 30045482
  • € 213.00 / 50µg
Description
Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fc gamma RI/CD64
Monoclonal
Unconjugated
P12314
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Medarex Patent Anti-Fc-Gamma-R1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Fc gamma R1
50 μg
CD Markers, Immunology
2209