missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RI/CD64 Antibody (OTI1A8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70702AF532
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Fc gamma RI/CD64 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FCGR1A | |
| Full length recombinant protein of human Fc gamma RI/CD64 (NP_000557) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1A8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| Mouse | |
| 0.1 mL | |
| 2209 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction