Learn More
Fc gamma RI/CD64 Antibody (OTI1A8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-45625
Product Code. 18749643
  • € 625.00 / 0.10mL
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry
OTI1A8