Learn More
Fc gamma RII/CD32 Antibody (AT10) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08998-0.2mg
 View more versions of this product
Product Code. 18628407
  • € 641.00 / 0.20mg
Description
Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fc gamma RII/CD32
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry
AT10
Unconjugated
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Human CD32.
0.2 mg
Immunology
2213
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG κ