missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/CD32 Antibody (AT10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65338V
This item is not returnable.
View return policy
Description
Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human, Porcine, Canine, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Fc gamma RII/CD32 | |
| Monoclonal | |
| DyLight 405 | |
| P12318 | |
| FCGR2B | |
| K562 cell line | |
| 0.1 mL | |
| Immunology | |
| 2213 | |
| Human, Pig, Canine, Primate | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| AT10 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB100-65338 recognizes the human CD32 antigen, a 40kD glycoprotein that acts as a low affinity receptor for IgG (also known as Fc gamma RII). The antigen mediates several functions including endocytosis, activation of secretion, cytotoxicity and immunomodulation. CD32 is expressed by B cells, monocytes, granulocytes and platelets. Clone AT10 blocks the binding of IgG to Fc gamma RII. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction