missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/CD32 Antibody (MM0270- 3L1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12161AF488
This item is not returnable.
View return policy
Description
Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Fc gamma RII/CD32 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| P31994 | |
| FCGR2B | |
| Human recombinant Fc gamma RII/CD32 (extracellular domain) | |
| 0.1 mL | |
| Immunology | |
| 2213 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| MM0270- 3L1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects human Fc gamma RIIa (CD32a) and Fc gamma RIIb (CD32b) | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction