missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/RIII (CD32/CD16) Antibody (190909), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1460M
This item is not returnable.
View return policy
Description
Fc gamma RII/RIII (CD32/CD16) Monoclonal antibody specifically detects Fc gamma RII/RIII (CD32/CD16) in Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Fc gamma RII/RIII (CD32/CD16) | |
| Monoclonal | |
| DyLight 594 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2214 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry | |
| 190909 | |
| 50mM Sodium Borate | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIIB, Thr30-Arg207, Accession # P08101 | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction