missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/RIII (CD32/CD16) Antibody (2.4G2), PE/Cy7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52644PECY7
This item is not returnable.
View return policy
Description
Fc gamma RII/RIII (CD32/CD16) Monoclonal antibody specifically detects Fc gamma RII/RIII (CD32/CD16) in Mouse samples. It is validated for Flow Cytometry
Specifications
| Fc gamma RII/RIII (CD32/CD16) | |
| Monoclonal | |
| PE-Cyanine7 | |
| CD16, CD16A, CD16-II, FCG3, FCGR3, FCGR3A, FCGRIII, FcGRIIIA, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20, Low affinity immunoglobulin gamma Fc region receptor III-A | |
| Mouse BALB/c Macrophage J774 Cell Line. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 2214 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Flow Cytometry | |
| 2.4G2 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction