missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIIA/CD32a Antibody (7.3), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
€ 541.00 - € 584.00
Specifications
| Antigen | Fc gamma RIIA/CD32a |
|---|---|
| Clone | 7.3 |
| Applications | Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689235
|
Novus Biologicals
NBP2-47830B-100ul |
100 μg |
€ 584.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678805
|
Novus Biologicals
NBP2-47830B |
0.1 mL |
€ 541.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIIA/CD32a Monoclonal antibody specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| Fc gamma RIIA/CD32a | |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Immunology | |
| PBS | |
| 2212 | |
| IgG1 κ | |
| Protein A or G purified |
| 7.3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a | |
| K562 and FcgRII+L cells | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title