Learn More
Fc gamma RIIA/CD32a Antibody (8.7), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47831MFV610
Product Code. 18783002
  • € 804.00 / 0.10mL
Description
Fc gamma RIIA/CD32a Monoclonal antibody specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fc gamma RIIA/CD32a
8.7
Flow Cytometry
Immunology
Monoclonal
mFluor Violet 610 SE
RUO
Store at 4C in the dark.