missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIII (CD16) Antibody (275003) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB19601JF525
This item is not returnable.
View return policy
Description
Fc gamma RIII (CD16) Monoclonal antibody specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Fc gamma RIII (CD16) | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII, Ala31-Thr215, Accession # Q5D5J5 | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| 2214 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 275003 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction