missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIII (CD16) Antibody (MEM-154), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-378G
This item is not returnable.
View return policy
Description
Fc gamma RIII (CD16) Monoclonal antibody specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Cell Surface), CyTOF
Specifications
| Fc gamma RIII (CD16) | |
| Monoclonal | |
| DyLight 488 | |
| CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
| Human granulocytes | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| 2214 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunoprecipitation, Flow Cytometry (Cell Surface), CyTOF | |
| MEM-154 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction