missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIIIA/CD16a Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98147CL1
This item is not returnable.
View return policy
Description
Fc gamma RIIIA/CD16a Polyclonal antibody specifically detects Fc gamma RIIIA/CD16a in Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| Fc gamma RIIIA/CD16a | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
| Produced in rabbits immunized with purified, recombinant Rat Fc gamma RIIIA/CD16a (Met 1-Pro 200; Accession#: ABO70343.1). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| 2214 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction