missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA2 Antibody (OTI3C7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72894B
This item is not returnable.
View return policy
Description
NCOA2 Monoclonal specifically detects NCOA2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NCOA2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| NCOA2 | |
| Recombinant protein fragment corresponding to amino acids 1-280 of human NCOA2 (NP_006531) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI3C7 | |
| Western Blot | |
| BHLHE75, bHLHe75MGC138808, Class E basic helix-loop-helix protein 75, EC 2.3.1.48, GRIP1glucocorticoid receptor-interacting protein-1, KAT13C, NCoA-2SRC2, nuclear receptor coactivator 2, TIF2hTIF2, Transcriptional intermediary factor 2 | |
| Mouse | |
| Affinity Purified | |
| Cancer, DNA Repair, Transcription Factors and Regulators | |
| 10499.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction