missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AU1 Epitope Tag Antibody, Alexa Fluor™ 350, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-452AF350
This item is not returnable.
View return policy
Description
AU1 Epitope Tag Polyclonal antibody specifically detects AU1 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| AU1 Epitope Tag | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Purified | |
| Epitope Tag |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 350 | |
| AU1 epitope tag, DTYRYI epitope tag, DTYRYI tag | |
| Goats were immunized with AU1 (DTYRYI) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
| 0.1 mL | |
| Epitope Tags | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction