missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FcεR1α Armenian Hamster anti-Mouse, BV786, Clone: MAR-1 (also known as MAR1), BD OptiBuild™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 751761
This item is not returnable.
View return policy
Description
FcεR1α Monoclonal antibody specifically detects FcεR1α in Mouse samples. It is validated for Flow Cytometry.
Specifications
| FcεR1α | |
| Monoclonal | |
| 0.2 mg/mL | |
| Ms FceR1a BV786 MAR-1 50 μg | |
| 1 x 50 μg | |
| Primary | |
| Mouse | |
| IgG |
| Flow Cytometry | |
| MAR-1 (also known as MAR1) | |
| Brilliant Violet 786 | |
| Armenian Hamster | |
| RUO | |
| FceR1a | |
| Refrigerate (2 to 8°C) | |
| Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction