missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FcεR1α Mouse anti-Human, BV750, Clone: AER-37 (also known as CRA-1, CRA1), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747788
This item is not returnable.
View return policy
Description
FcεR1α Monoclonal antibody specifically detects FcεR1α in Human samples. It is validated for Flow Cytometry.
Specifications
| FcεR1α | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| AER-37 (also known as CRA-1, CRA1) | |
| Brilliant Violet 750 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction