missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCAR/CD89 Antibody (013), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90841MFV450
This item is not returnable.
View return policy
Description
FCAR/CD89 Monoclonal antibody specifically detects FCAR/CD89 in Rat samples. It is validated for ELISA
Specifications
| FCAR/CD89 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| ELISA | |
| 013 | |
| ELISA | |
| CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Rat FCAR/CD89 (Accession#: NP_973721.1; Met1-Asn228). | |
| 0.1 mL | |
| Immunology | |
| 2204 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction