missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCAR/CD89 Antibody (14A8 (14.1)), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20161MFV610
This item is not returnable.
View return policy
Description
FCAR/CD89 Monoclonal antibody specifically detects FCAR/CD89 in Human samples. It is validated for Flow Cytometry
Specifications
| FCAR/CD89 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 14A8 (14.1) | |
| Flow Cytometry | |
| CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
| The original antibody was generated by immunizing HCO12 trangene containing mice with recombinant soluble CD89 antigen. | |
| 0.1 mL | |
| Immunology | |
| 2204 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction