missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FCAR Recombinant Rabbit Monoclonal Antibody (013)
GREENER_CHOICE

Product Code. 16314744
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16314744 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16314744 Supplier Invitrogen™ Supplier No. MA531293

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Rat CD89/FCAR.

This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCAR
Applications ELISA
Classification Recombinant Monoclonal
Clone 013
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Fcar
Gene Accession No. 0
Gene Alias CD89; CTB-61M7.2; Fc alpha receptor; Fc fragment of IgA receptor; Fc fragment of IgA, receptor for; FcalphaRI; FCAR; IgA Fc receptor; immunoglobulin A Fc receptor; immunoglobulin A Fc receptor transcript variant 1; immunoglobulin A Fc receptor transcript variant 3; immunoglobulin A Fc receptor transcript variant 7; immunoglobulin A Fc receptor transcript variant 9; immunoglobulin A Fc receptor transcript variant deltaEC1; immunoglobulin A Fc receptor transcript variant deltaEC1 delta TM/C; immunoglobulin alpha Fc receptor
Gene Symbols Fcar
Host Species Rabbit
Immunogen Recombinant Rat CD89/FCAR protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 365183
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.