Learn More
Invitrogen™ FceR1 alpha Monoclonal Antibody (CRA2 (AER24))
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA14998
Product Code. 11574333
  • € 764.00 / 100µg
Description
MA1-4998 detects Fc epsilonR1 alpha from human samples. The CRA2 (AER24) monoclonal antibody reacts with the Fc epsilonR1 alpha subunit on a region that overlaps the region of the IgE binding site, thus it competes with IgE for the receptor binding. Since MA1-4998 has been successfully used in Western blot, flow cytometry, and immunohistochemistry applications. The MA1-4998 immunogen is region of Fc epsilonR1 alpha which overlaps with the IgE binding site. The epitope has been mapped to amino acids 110-197.
Fc epsilon RI alpha, also known as FceR1 alpha, is a subunit of the high-affinity receptor for IgE, primarily expressed on mast cells and basophils. Its expression is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits, which are essential for signal transduction. The beta and gamma subunits contain immunoreceptor tyrosine-based activation motifs (ITAMs). The Fc epsilon RI complex plays a crucial role in triggering IgE-mediated allergic reactions. When allergen-bound IgE molecules bring together two or more high-affinity IgE receptors, it leads to the release of mediators such as histamine, which are responsible for allergy symptoms. This process couples allergens and mast cells to initiate inflammatory responses characteristic of allergic disorders like hay fever and asthma. The release of histamine and proteases also leads to the synthesis of prostaglandins and leukotrienes, which are potent effectors of the hypersensitivity response.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FceR1 alpha
Monoclonal
1 mg/mL
PBS with 50% glycerol and no preservative; pH 7.4
P12319
FCER1A
Recombinant extracellular portion of recombinant human FcER1 alpha (corresponds to amino acids Met-26-197 excluding signal peptide 1-25).
100 μg
Primary
Human
Antibody
IgG1 κ
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
CRA2 (AER24)
Unconjugated
FCER1A
alpha polypeptide; fc epsilon r1; Fc epsilon receptor Ia; Fc epsilon RI alpha-chain; Fc fragment of IgE receptor Ia; Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide; Fc IgE receptor, alpha polypeptide; Fc receptor, IgE, high affinity I, alpha polypeptide; Fce1a; Fc-epsilon RI-alpha; fcepsilonri; FCER1A; FcERI; Fcr-5; high affinity immunoglobulin epsilon receptor alpha-subunit; high affinity immunoglobulin epsilon receptor subunit alpha; IgE Fc receptor subunit alpha; Iger01; immunoglobulin E receptor, high-affinity, of mast cells, alpha polypeptide; RATIGER01; RP11-550P17.3
Mouse
IgG fraction
RUO
2205
-20°C, Avoid Freeze/Thaw Cycles