missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCGR1A Mouse anti-Human, Clone: OTI3D3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506343S
This item is not returnable.
View return policy
Description
CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.Specifications
| FCGR1A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P12314 | |
| FCGR1A | |
| Full length human recombit protein of human FCGR1A produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D3 | |
| Unconjugated | |
| FCGR1A | |
| CD64, CD64A, FCRI, IGFR1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2209 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction