Learn More
FcgR4/CD16-2 Antibody (011), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-05889MFV450
This item is not returnable.
View return policy
Description
FcgR4/CD16-2 Monoclonal antibody specifically detects FcgR4/CD16-2 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| FcgR4/CD16-2 | |
| RUO | |
| Mouse |
| Immunohistochemistry (Paraffin) | |
| Primary | |
| IgG |