missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FcgR4/CD16-2 Antibody (012), Alexa Fluor™ 647 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-05890AF647
This item is not returnable.
View return policy
Description
FcgR4/CD16-2 Monoclonal antibody specifically detects FcgR4/CD16-2 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| FcgR4/CD16-2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 246256 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 012 | |
| 50mM Sodium Borate | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FcgR4/CD16-2 (NP_653142.2; Met 1-Gln 203). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction