missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCHSD1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41137AF647
This item is not returnable.
View return policy
Description
FCHSD1 Polyclonal antibody specifically detects FCHSD1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| FCHSD1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| FCH and double SH3 domains 1, FCH and double SH3 domains protein 1, FLJ00007, nervous wreck homolog 2, NWK2 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human FCHSD1. The immunogen is located within the last 50 amino acids of FCHSD1. Amino Acid Squence: PPAKAPDPGHPDPLT | |
| 0.1 mL | |
| Neuroscience | |
| 89848 | |
| Store at 4°C in the dark. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction