missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FCHSD1 Polyclonal specifically detects FCHSD1 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | FCHSD1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Alias | FCH and double SH3 domains 1, FCH and double SH3 domains protein 1, FLJ00007, nervous wreck homolog 2, NWK2 |
| Gene Symbols | FCHSD1 |
| Host Species | Chicken |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?