missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRL2/FcRH2 Antibody (296902), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2048M
This item is not returnable.
View return policy
Description
FCRL2/FcRH2 Monoclonal antibody specifically detects FCRL2/FcRH2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| FCRL2/FcRH2 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79368 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 296902 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD307b antigen, Fc receptor homolog 2, Fc receptor-like 2, FcRH2, FCRH2SPAP1B, FcRL2, FcR-like protein 2, IFGP family protein 4, IFGP4, Immunoglobulin receptor translocation-associated protein 4, immunoglobulin superfamily Fc receptor, gp42, IRTA4SPAP1C, SH2 domain containing phosphatase anchor protein 1, SH2 domain-containing phosphatase anchor protein 1Fc receptor-like protein 2, SPAP1A, SPAP1immune receptor translocation-associated protein 4 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL2/FcRH2, Glu15-Asp395, Accession # Q96LA5 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction