missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRL3/FcRH3 Antibody (546828), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3126MFV500
This item is not returnable.
View return policy
Description
FCRL3/FcRH3 Monoclonal antibody specifically detects FCRL3/FcRH3 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| FCRL3/FcRH3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 546828 | |
| Flow Cytometry, CyTOF-ready | |
| CD307c, Fc receptor homolog 3, Fc receptor-like 3, Fc receptor-like protein 3, FCRH3, fcR-like protein 3, hIFGP3, IFGP3, immunoglobulin superfamily receptor translocation associated protein 3, IRTA3, SPAP2 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL3/FcRH3, Arg14-Arg569, Accession # Q96P31 | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers | |
| 115352 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction